ContactUsLinkComponent
VWR® Low-Level Radiation Bags
  89026-660
Small Business Enterprise
undefined
VWR® Low-Level Radiation Bags
  89026-660
Small Business Enterprise
  • Thickness:
    2 mil.
  • Cat. No.:
    89026-660
  • W×L:
    965×1220 mm (38×48")
These polypropylene bags meet all federal and state regulations for the handling of low-level radiation waste.
Bags are transparent yellow and feature the universal radiation symbol.