ContactUsLinkComponent
Linearity FD Urine Chemistry Panel 2 for Abbott Systems, AUDIT Microcontrols
Linearity FD Urine Chemistry Panel 2 for Abbott Systems, AUDIT Microcontrols
Catalog # 77497-282
CAS Number:  
undefined
Linearity FD Urine Chemistry Panel 2 for Abbott Systems, AUDIT Microcontrols
Catalog # 77497-282
Supplier Number:  K898M-5
CAS Number:  
Specifications
  • Description:
    Linearity FD Urine Chemistry Panel 2 for Abbott Systems, 5 vials
  • Size:
    5x 3 ml
  • Cat. No.:
    77497-282
Specifications
About this item
Linearity FD Urine Chemistry Panel 2 for Abbott Systems is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, and Sodium.
  • Freeze Dried
The Linearity FD Urine Chemistry Panel 2 for Abbott Systems is assayed quality control material consisting of five levels of human serum.
Each level of the Linearity FD Urine Chemistry Panel 2 for Abbott Systems contains the following analytes: Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, and Sodium.
These five levels of each set demonstrate a linear relationship to each other for their respective analytes.
It is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, and Sodium.
Ordering information: Send PO to [email protected]