Specifications
- Description:Linearity FD Urine Chemistry Panel 1 for Abbott Systems, 10 vials
- Size:10x 3 ml
- Cat. No.:77497-280
Specifications
About this item
Linearity FD Urine Chemistry Panel 1 for Abbott Systems is intended to simulate human patient urine chemistry samples for purpose of determining linearity, calibration verification and verification of reportable range for Phosphorous, Urea Nitrogen and Uric Acid.
- Freeze Dried
The Linearity FD Urine Chemistry Panel 1 for Abbott Systems is assayed quality control material consisting of two sets of five levels of aqueous buffer.
Each level of the Linearity FD Urine Chemistry Panel 1 for Abbott Systems Set 1 contains the following analytes: Phosphorous, Urea Nitrogen.
Each level of the Linearity FD Urine Chemistry Panel 1 for Abbott Systems Set 2 contains the following analyte: Uric Acid.
These five levels of each set demonstrate a linear relationship to each other for their respective analytes.
It is intended to simulate human patient urine chemistry samples for purpose of determining linearity, calibration verification and verification of reportable range for Phosphorous, Urea Nitrogen and Uric Acid.