VWR® Autoclavable Biohazard Bags and Containers
About this item
Double-thick bags are ideal for sterilizing animal cages and other objects with sharp corners.
- High-integrity polypropylene
- Available with temperature indicator
- Resists punctures, tears, and leaks
Ideal for processing biohazardous materials that require autoclaving prior to disposal.
Twist-tie closure allows bag to expand and vent steam during cooling. Bags resist punctures, tears, and leaks and meet the 165 g dart drop standard performed as required by ASTM D1709-98. Each bag with indicator is imprinted with the universal biohazard symbol.
Indicator bags have the words 'Contents Have Been Autoclaved' printed in a chemically active ink. Before autoclaving, the ink is white. After autoclaving is complete, it darkens, providing instant proof that biohazardous waste has been autoclaved.
Certifications: Meets the 165 g dart drop standard (ASTM D1709-98).
1 Options Available Below
Product Details & Documents
Double-thick bags are ideal for sterilizing animal cages and other objects with sharp corners.
- High-integrity polypropylene
- Available with temperature indicator
- Resists punctures, tears, and leaks
Ideal for processing biohazardous materials that require autoclaving prior to disposal.
Twist-tie closure allows bag to expand and vent steam during cooling. Bags resist punctures, tears, and leaks and meet the 165 g dart drop standard performed as required by ASTM D1709-98. Each bag with indicator is imprinted with the universal biohazard symbol.
Indicator bags have the words 'Contents Have Been Autoclaved' printed in a chemically active ink. Before autoclaving, the ink is white. After autoclaving is complete, it darkens, providing instant proof that biohazardous waste has been autoclaved.
Certifications: Meets the 165 g dart drop standard (ASTM D1709-98).
Product Family Options
Orange Bags with Indicator
Product Information
- CapacityFitsW×LThicknessAvailabilityPrice
- 203×305 mm (8×12")2 mil.Discontinued-
- 356×483 mm (14×19")2 mil.Discontinued-
- 635×889 mm (25×35")2 mil.Discontinued-
- 914×1143 mm (36×45")2 mil.Discontinued-
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



