Order Entry
United States
Orders LinkContactUsLinkComponent
Compartmental Bubble Pouch, Com-Pac

Compartmental Bubble Pouch, Com-Pac

350×150 mm (13.75×6")
Catalog # 11217-558
Supplier:   Com-Pac
Supplier #:  BLP-1000
Compartmental Bubble Pouch, Com-Pac
Catalog # 11217-558
Supplier:  Com-Pac
Compartmental Bubble Pouch, Com-Pac
Compartmental Bubble Pouch, Com-Pac

350×150 mm (13.75×6")

Price
$63.79
Log in for availability
Case of 25
Product Details & Documents

Economical pouch features six 5 cm (2") test tube-sized compartments to keep test tubes separated.

Protective cushioning material reduces risk of breakage.

Specifications
  • W×L:
    350×150 mm (13.75×6")
  • Cat. No.:
    11217-558
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.