Order Entry
United States
Orders LinkContactUsLinkComponent
 

3-Pentanol ≥98.0%

3-Pentanol ≥98.0%
Supplier:  TCI America
Certificates
3-Pentanol ≥98.0%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 584-02-1 MDL Number: MFCD00004574 Molecular Formula: C5H12O Molecular Weight: 88.15 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 116 Melting point (°C): -8 Flash Point (°C): 34 Specific Gravity (20/20): 0.82

Other Product Names / Synonyms: Diethylcarbinol

  • CAS Number:584-02-1
  • Formula:C₅H₁₂O
  • MDL Number:MFCD00004574
  • Molecular Weight:88.15 g/mol
  • Merck Index:13,07197
  • Boiling Pt:116 °C
  • Melting Pt:–8 °C
  • Density:0.82 (20 °C)
  • Flash Pt:34 °C
  • Storage Temperature:Ambient
  • EINECS:209-526-7
  • UN:1105
  • ADR:3,III
Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    98.0% (GC)
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.