Order Entry
United States
ContactUsLinkComponent
Phenol liquified ≥89.0% stabilized USP, Macron Fine Chemicals™
Phenol liquified ≥89.0% stabilized USP, Macron Fine Chemicals™
Catalog # MK061006
CAS Number:  
undefined
Phenol liquified ≥89.0% stabilized USP, Macron Fine Chemicals™
Catalog # MK061006
Supplier Number:  0610-06
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    2.5 kg
  • Cat. No.:
    MK061006
  • Supplier no.:
    0610-06

Specifications

Specification Test Results

  • USP - Assay (C₆H₅OH):
    ≥89.0 %
  • USP - Benzene:
    ≤2.0 ppm
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Clarity of Solution and Reaction:
    Passes Test
  • USP - Nonvolatile Residue:
    ≤0.05 %
  • Distilling Range - Final Temperature:
    ≤182.5 °C
  • Stabilizer:
    Passes Test
  • JP - Identification (1):
    Passes Test
  • JP - Identification (2):
    Passes Test
  • JP - Boiling Point:
    ≤182 °C
  • JP - Clarity and Color of Solution and Acidity or Alkalinity:
    Passes Test
  • JP - Residue on Evaporation:
    ≤0.05 %
  • JP - Assay:
    ≥88.0 %

Specification Test Results

About this item

 :
 :94.11 g/mol
Merck Index:12,07390
Boiling Pt:181.7 °C (1013 hPa)
Melting Pt:40.8 °C
Density:1.0722 g/cm³ (20 °C)
Flash Pt:81 °C
ADR:6.1,II
 :Management of Change (MOC) category = R
 :Stabilised with citric acid 20 ppm
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
REACH:01-2119471329-32
 :GHS06
 :GHS05
 :GHS08
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143