Phenol red, BAKER ANALYZED® ACS indicator, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:143-74-8
:
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552
Merck Index:13,07329
Melting Pt:>300 °C
:Management of Change (MOC) category = N
:354.38 g/mol
Storage Temperature:Ambient
EINECS:205-609-7
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Clarity of Solution:Passes Test
- Visual Transition Interval - pH…6.8 (Yellow):Passes Test
- Visual Transition Interval - pH…8.2 (Red):Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot NumberBrand Definition Baker Analyzed
Datasheet
T254 Product Specifications
Additional Product Details
Frequently Bought Together