Order Entry
United States
ContactUsLinkComponent
Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Catalog # MK1674-21
CAS Number:  
undefined
Tannic acid ≥96.0% (dry basis) FCC, Macron Fine Chemicals™
Catalog # MK1674-21
Supplier Number:  MACR1674-21
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Box for dry chemical with liner
  • Size:
    25 kg
  • Cat. No.:
    MK1674-21

Specifications

Specification Test Results

  • Assay () (dried basis) (FCC):
    ≥96.0 %
  • Appearance (fluffy, light tan to brown powder):
    Passes Test
  • Solubility:
    Passes Test
  • Gums or Dextrin:
    Passes Test
  • Resinous Substances:
    Passes Test
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Loss on Drying at 105°C:
    ≤7.0 %
  • Residue on Ignition:
    ≤1.0 %
  • Lead (Pb):
    ≤2 mg/kg

Specification Test Results

About this item

 :
 :1701.2 g/mol
Merck Index:12,09221
Melting Pt:218…219 °C
Density:>1 g/cm³ (20 °C)
 :Management of Change (MOC) category = RL
Flash Pt:199 °C
EINECS:215-753-2
CAS Number:1401-55-4
 :85085-33-0
Formula:C₇₆H₅₂O₄₆
MDL Number:MFCD00066397