


Tetrahydrofuran, low water content ≥99.8% (by GC, corrected for water content) contains no preservatives for HPLC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
:Management of Change (MOC) category = N
Flash Pt:–21.5 °C
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance (Clear, colorless liquid):Passes Test
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-320 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.030
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.15
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:≤0.40
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤212
- Assay (by GC, corrected for water):≥99.8 %
- Color (APHA):≤10
- Peroxide (as H₂O₂) (at time of fill):≤10 ppm
- Residue after Evaporation:≤2.0 ppm
- Water (by KF, coulometric):≤50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number9439 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together