




Pyridine, low water content ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:110-86-1
:
Formula:C₅H₅N
MDL Number:MFCD00011732
Merck Index:13,08060
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
:Management of Change (MOC) category = N
:79.1 g/mol
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
Specification Test Results
- Assay (by GC, corrected for water):≥99.0 %
- Ammonia (as NH₃):≤0.002 %
- Color (APHA):≤10
- Reducing Substances:Passes Test
- Residue after Evaporation:≤5 ppm
- Solubility in H₂O:Passes Test
- Sulfate (SO₄):≤0.001 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 375 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 nm:≤1.00
- Water (by KF, volumetric):≤0.01 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - Copper (Cu):≤5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number9393 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor J.T.Baker® LCMS and HPLC Solvents and Reagents Avantor Portfolio of Chemicals
Product Literature
spec_JT9393-2
Product specification
Frequently Bought Together