Order Entry
United States
ContactUsLinkComponent
 
Tin ≥99.9%, granular, BAKER ANALYZED® ACS 20 mesh, J.T.Baker®
Catalog # JT4150-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Tin ≥99.9%, granular, BAKER ANALYZED® ACS 20 mesh, J.T.Baker®
Catalog # JT4150-1
Supplier Number:  4150-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    Poly bottle
  • Size:
    500 g
  • Cat. No.:
    JT4150-1
  • Supplier no.:
    4150-01

Specifications

Specification Test Results

  • ACS - Assay (Sn) (by EDTA titrn):
    ≥99.5 %
  • ACS - Antimony (Sb)(by AAS):
    ≤0.02 %
  • ACS - Copper (Cu)(by AAS):
    ≤0.005 %
  • ACS - Iron (Fe)(by AAS):
    ≤0.01 %
  • ACS - Lead (Pb)(by AAS):
    ≤0.005 %
  • Trace Impurities - Arsenic (As):
    ≤1 ppm
  • Mesh - On U.S. No. 18 Sieve:
    ≤5 %
  • Mesh - Thru U.S. No. 35 Sieve:
    ≤30 %

Specification Test Results

About this item

 :
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.31 g/cm³ (25 °C)
 :Management of Change (MOC) category = N
 :118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8
CAS Number:7440-31-5
Formula:Sn
MDL Number:MFCD00133862