

Tin(II) chloride dihydrate 98.0-103.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:10025-69-1
:
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863
:225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Assay (SnCl₂·2H₂O):98.0 - 103.0 %
- Solubility in HCl:Passes Test
- Sulfate (SO₄):Passes Test
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.003 %
- Lead (Pb):≤0.01 %
- Magnesium (Mg):≤0.01 %
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.010 %
- Trace Impurities - Arsenic (As):≤2.000 ppm
- Trace Impurities - Mercury (Hg):≤0.050 ppm
Specification Test Results
Related Information
Frequently Bought Together