


Sulfuric acid 10.0 N in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # MKH37905
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:PE bottle, narrow mouth
- Size:1 L
- Cat. No.:MKH37905
- Supplier no.:H379-05
Specifications
Specification Test Results
- Normality:9.95 - 10.05
Specification Test Results
About this item
:
:98.08
ADR:8,II
:Management of Change (MOC) category = N
EINECS:231-639-5
UN:2796
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589