About this item
Locking incubators with digital lock, digital temperature and timer controls.
- Accurate temperature control with easy to use digital control panel
- Durable cold rolled epoxy painted steel construction
- Insulated glass viewing window for improved visibility
- Built-in digital timer of 0 - 9999 minutes for managing timed processes
- Digital keypad lock for securing samples without the need for a key
- Choice of 2 different sizes: 30 or 65 L capacity
Large handle and viewing window. The incubators have a stainless steel interior. The shelf load is rated at 39 kg for the 30 L units and 46 kg for the 65 L incubators.
These locking incubators features a tubular heating element, controlled by accurate PID and feedback from a PT100 sensor. Temperature fluctuation is ±0.2 °C, and temperature uniformity is ±2.0 °C of the setpoint.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76644-936
- 76644-938
- Temperature Fluctuation (time):
- ±0.2 °C
- ±0.2 °C
- Temperature Variation (spatial):
- ±0.2 °C
- ±0.2 °C