Order Entry
United States
ContactUsLinkComponent
 
Thermo Scientific™ Bioreactor Tubes, 50 ml
Catalog # 77392-120
Thermo Scientific™ Bioreactor Tubes, 50 ml
Catalog # 77392-120
Supplier Number:  332260

Specifications

  • Sterility:
    Sterile
  • Capacity:
    50 ml
  • Description:
    Bioreactor tubes, concial
  • Material:
    PP
  • Environmentally Preferable:
  • Cat. No.:
    77392-120
  • Packaging:
    Case of 90

Specifications

About this item

Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition.

  • Polypropylene tube
  • HDPE cap
  • 2 µm PTFE filter membrane
  • Polystyrene dial
  • Suitable for suspension culture
  • Sterile - SAL 10⁻⁶
  • Non-pyrogenic
  • Non-cytotoxic
  • Shelf life two years

Ideal for screening, small volume high throughput analysis. Protein expression suspension culture volumes ranging from 10 to 25 ml. Convenient pack sizes. Four different sizes of ventilation (dial type) on the cap.

Packaging: 3 per pack; 30 packs per case.