


Potassium chromate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:7789-00-6
:
Formula:K₂CrO₄
MDL Number:MFCD00011368
:194.19 g/mol
Merck Index:12,07784
Boiling Pt:1000 °C (1013 hPa)
Melting Pt:985 °C
Density:2.73 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3087
ADR:5.1,III
:Management of Change (MOC) category = N
EINECS:232-140-5
:GHS08
:GHS07
:GHS09
Specification Test Results
- Assay (K₂CrO₄):≥99.0 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:8.6 - 9.8
- Chloride (Cl):≤0.005 %
- Sulfate (SO₄):≤0.03 %
- Calcium (Ca):≤0.005 %
- Sodium (Na)(by FES):≤0.02 %
Specification Test Results
Related Information
Certificates
Search by Lot Number3058 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
Frequently Bought Together