

Tetrasodium diphosphate decahydrate 99.0-103.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:13472-36-1
:
Formula:Na₄P₂O₇·10H₂O
MDL Number:MFCD00149200
:446.06 g/mol
Melting Pt:80 °C
Density:1.82 g/cm³ (25 °C)
:Management of Change (MOC) category = N
EINECS:231-767-1
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (Na₄P₂O₇·10H₂O):99.0 - 103.0 %
- Insoluble Matter:≤0.01 %
- pH of 5% Solution at 25°C:9.5 - 10.5
- Chloride (Cl):≤0.002 %
- Nitrogen Compounds (as N):≤0.001 %
- Sulfate (SO₄):≤0.005 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
Specification Test Results
Related Information
Certificates
Search by Lot Number3850 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
Frequently Bought Together