Technic™ Flock-Lined Neoprene-Over-Natural-Rubber Gloves,
Supplier: MAPA
Certificates
About this item
Gloves offer reliable hand protection for chemical processing, metal finishing, mechanical work, painting, degreasing, plating, bleaching, leather tanning, commercial dish/pot washing, janitorial, and general maintenance applications.
- Color: black
- Comfortable flock lining
- Gloves are chemical-resistant and heavy-duty
Flexible, comfortable, hand-specific design features a flat cuff and curved fingers for a natural fit. Embossed diamond grip for safe, easy handling of wet and greasy objects.
Warning: This product contains natural rubber latex and is not recommended for those who are allergic to natural rubber proteins.
Certifications: In compliance with 21 CFR 170-199. CE marked.
Specifications
- Catalog No:
- 32917-231
- 32917-232
- 32917-229
- 32917-230
- 32917-224
- 32917-242
- 32917-223
- 32917-220
- 32917-240
- 32917-241
- 32917-233
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex