AlphaTec® 29-865 Unsupported Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Type 29-865 neoprene gloves offer broad-spectrum protection with excellent resistance to a wide range of chemicals, including oils, acids, caustics, and solvents.
- Hand-specific
- Straight cuffs
- Sand patch finish
- Case-hardened for increased resistance
Flock-lined gloves are case-hardened to increase chemical and abrasion resistance over ordinary neoprene gloves. Gloves are suitable for petrochemicals and refining operations, cleaning and degreasing, and electronics applications.
Certifications: Complies with FDA food handling requirements 21 CFR (USA) and European Directives.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.
Specifications
- Catalog No:
- 32890-052
- 32890-058
- 32890-060
- 32890-054
- 32890-056
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- EN Norm:
- 3111
- 3111
- 3111
- 3111
- 3111