Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (TRITC)
Supplier: AAT BIOQUEST INC
Certificates
About this item
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: TRITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 77022-344
- 77021-640
- Antigen Symbol:
- CD4
- CD4
- Antigen Name:
- CD4 antigen
- CD4 antigen
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- TRITC (Tetramethylrhodamine Isothiocyanate)
- TRITC (Tetramethylrhodamine Isothiocyanate)
- Clone:
- RPA-T4
- RPA-T4
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Purification:
- Affinity purification
- Affinity purification
- Storage Temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping Temperature:
- 20
- 20
Specifications
Frequently Bought Together