Order Entry
United States
ContactUsLinkComponent
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC/iFluor® 750)
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC/iFluor® 750)
Supplier:  AAT BIOQUEST INC
Certificates
undefined
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC/iFluor® 750)
Supplier:  AAT BIOQUEST INC

Specifications

  • Catalog No:
  • 77021-348
  • 77025-052
  • 77021-354
  • Antigen Symbol:
  • CD3e
  • CD3e
  • CD3e
  • Antigen Name:
  • CD3e antigen
  • CD3e antigen
  • CD3e antigen
  • Antibody Type:
  • Secondary
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/iFluor® 750
  • APC (Allophycocyanin)/iFluor® 750
  • APC (Allophycocyanin)/iFluor® 750
  • Clone:
  • SK7
  • SK7
  • SK7
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Purification:
  • Affinity purification
  • Affinity purification
  • Affinity purification
  • Storage Temperature:
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Shipping Temperature:
  • 20
  • 20
  • 20

Specifications

Frequently Bought Together