Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Cat. No.:95056-982
- Suture size:4 - 0
- Suture length:45 cm (18")
- Suture material:Green braided monofilament, 12-strand
- Supplier no.:X183H
Nonabsorbable, braided, sterile, surgical sutures are composed of polyethylene terephthalate
Prepared from fibers of high molecular weight, long-chain, linear polyesters having reccurent aromatic rings as an integral component. The highly adherent coating is a relatively nonreactive nonabsorbable compound which acts as a lubricant to mechanically improve the physical properties of the uncoated suture by improving handling qualities as contrasted to the braided, uncoated fiber. The suture is braided for optimal handling properties, and dyed green for enhanced visibility in the surgical field.