Order Entry
United States
ContactUsLinkComponent
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PE/iFluor® 700)
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PE/iFluor® 700)
Supplier:  AAT BIOQUEST INC
Certificates
undefined
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PE/iFluor® 700)
Supplier:  AAT BIOQUEST INC

Specifications

  • Catalog No:
  • 77023-034
  • 77021-936
  • 77021-632
  • Antigen Symbol:
  • CD161
  • CD161
  • CD161
  • Antigen Name:
  • CD161 antigen
  • CD161 antigen
  • CD161 antigen
  • Antibody Type:
  • Secondary
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/iFluor® 700
  • PE (Phycoerythrin)/iFluor® 700
  • PE (Phycoerythrin)/iFluor® 700
  • Clone:
  • HP-3G10
  • HP-3G10
  • HP-3G10
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Purification:
  • Affinity purification
  • Affinity purification
  • Affinity purification
  • Storage Temperature:
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Shipping Temperature:
  • 20
  • 20
  • 20

Specifications

Frequently Bought Together