Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Violet)
Supplier: AAT BIOQUEST INC
Certificates
About this item
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: mFluor™ Violet
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 77023-014
- 77023-724
- Antigen Symbol:
- CD161
- CD161
- Antigen Name:
- CD161 antigen
- CD161 antigen
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Violet
- mFluor™ Violet
- Clone:
- HP-3G10
- HP-3G10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage Temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping Temperature:
- 20
- 20
Specifications
Frequently Bought Together