Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 532)
Supplier: AAT BIOQUEST INC
Certificates
About this item
4H11 is an anti-human monoclonal antibody that recognizes the CD34 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 77023-792
- 77024-768
- Antigen Symbol:
- CD34
- CD34
- Antigen Name:
- CD34 antigen
- CD34 antigen
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 532
- iFluor® 532
- Clone:
- 4H11
- 4H11
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Purification:
- Affinity purification
- Affinity purification
- Storage Temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping Temperature:
- 20
- 20
Specifications
Frequently Bought Together