Order Entry
United States
ContactUsLinkComponent
 
Anti-CD19 Mouse Monoclonal Antibody (iFluor® 350) [clone: HIB19]
 
undefined
Anti-CD19 Mouse Monoclonal Antibody (iFluor® 350) [clone: HIB19]

 

  • Catalog No:
  • 77024-616
  • 77024-398
  • Antigen Symbol:
  • CD19
  • CD19
  • Antigen Name:
  • CD19 antigen
  • CD19 antigen
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® 350
  • iFluor® 350
  • Clone:
  • HIB19
  • HIB19
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage Temperature:
  • Refrigerated (2...8 °C); minimize light exposure
  • Refrigerated (2...8 °C); minimize light exposure
  • Shipping Temperature:
  • 20
  • 20

 

Frequently Bought Together