Anti-CD14 Mouse Monoclonal Antibody (PE/iFluor® 750) [clone: 61D3]
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
: For research use only.
Type: Secondary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: PE/iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- 77023-170
- 77022-978
- 77021-042
- Antigen Symbol:
- CD14
- CD14
- CD14
- Antigen Name:
- CD14 antigen
- CD14 antigen
- CD14 antigen
- Antibody Type:
- Secondary
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/iFluor® 750
- PE (Phycoerythrin)/iFluor® 750
- PE (Phycoerythrin)/iFluor® 750
- Clone:
- 61D3
- 61D3
- 61D3
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Affinity purification
- Storage Temperature:
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Shipping Temperature:
- 20
- 20
- 20
Frequently Bought Together