Order Entry
United States
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody (iFluor® 532) [clone: L17F12]
Anti-CD5 Mouse Monoclonal Antibody (iFluor® 532) [clone: L17F12]
Supplier:  AAT Bioquest
Certificates
undefined
Anti-CD5 Mouse Monoclonal Antibody (iFluor® 532) [clone: L17F12]
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • 77024-942
  • 77022-366
  • Antigen Symbol:
  • CD5
  • CD5
  • Antigen Name:
  • CD5 antigen
  • CD5 antigen
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® 532
  • iFluor® 532
  • Clone:
  • L17F12
  • L17F12
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage Temperature:
  • Refrigerated (2...8 °C); minimize light exposure
  • Refrigerated (2...8 °C); minimize light exposure
  • Shipping Temperature:
  • 20
  • 20

Specifications

Frequently Bought Together