Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]
Catalog # 77022-750
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Antibody Type:Primary
- Antigen Name:CD4 antigen
- Antigen Symbol:CD4
- Clonality:Monoclonal
- Clone:RPA-T4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Storage Temperature:Refrigerated (2...8 °C); minimize light exposure
- Shipping Temperature:20
- Purification:Affinity purification
- Cat. No.:77022-750
- Packaging:Vial
Specifications
About this item
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human