ContactUsLinkComponent
NEBNext® Primers for Epigenetics (Unique Dual Index Set 3)
NEBNext® Primers for Epigenetics (Unique Dual Index Set 3)
Catalog # 77441-302
CAS Number:  
undefined
NEBNext® Primers for Epigenetics (Unique Dual Index Set 3)
Catalog # 77441-302
Supplier Number:  E3404S
CAS Number:  
Specifications
  • Description:
    NEBNext® primers for epigenetics (Unique Dual Index Set 3)
  • Size:
    96 reactions
  • Cat. No.:
    77441-302
Specifications
About this item
The NEBNext Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries.
  • Optimized for NEBNext Epigenetics workflows
This 96 reaction kit contains premixed i5 and i7 8-base index primers, and each well of a 96-well plate contains an i5 and i7 primer pair.