NEBNext® Primers for Epigenetics (Unique Dual Index Set 2B)
Catalog # 77441-300
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:NEBNext® primers for epigenetics (Unique Dual Index Set 2B)
- Size:24 reactions
- Cat. No.:77441-300
Specifications
About this item
The NEBNext® Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries.
- Optimized for NEBNext Epigenetics workflows
This 24-reaction kit is supplied in a plate with premixed i5 and i7 8-base index primers in 24 wells.