


Mercury(II) nitrate 0.0250 Normal (N/40) titrant
Catalog # RC4715-32
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Glass bottle, amber
- Size:1 L
- Cat. No.:RC4715-32
- Supplier no.:R4715000-1C
Specifications
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):0.02500 ± 0.00001 N at 20.0 °C
Specification Test Results
About this item
:
:324.6
Melting Pt:79 °C
Density:4.3 g/cm³ (20 °C)
EINECS:233-152-3
UN:1625
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:10045-94-0
Formula:HgN₂O₆
MDL Number:MFCD00011038