

Methyl ethyl ketone ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
: Avantor
:78-93-3
:
:C₂H₅COCH₃
:MFCD00011648
:13,06097
:79.6 °C (1013 hPa)
:–86 °C
:0.8054 g/cm³ (20 °C)
:–4 °C
:201-159-0
:1193
:3,II
:Management of Change (MOC) category = N
:72.11 g/mol
:01-2119457290-43
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended by default and after discussion
- ACS - Assay (CH₃COCH₂CH₃) (by GC, corrected for water):≥99.0 %
- ACS - Color (APHA):≤15
- ACS - Residue after Evaporation:≤0.0025 %
- ACS - Titrable Acid (meq/g):≤0.0005
- ACS - Water (by KF, volumetric):≤0.20 %
Frequently Bought Together