Paraffin Oil light, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:8012-95-1
:
Merck Index:13,07093
Boiling Pt:300…500 °C (1013 hPa)
Density:0.86 g/cm³ (20 °C)
Flash Pt:215 °C (closed cup)
EINECS:232-384-2
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Color (APHA):20 max
- Description:To pass test
- Infrared spectrum:Conforms to standard
- Limit of polynuclear compounds:To pass test
- Neutrality:To pass test
- Readily carbonizable substances:To pass test
- Solid paraffin:To pass test
- Solubility:To pass test
- Specific Gravity at 25/25°C:0.845-0.875
- Transmittance at 420 nm:93%T min
Specification Test Results
Frequently Bought Together