
Methylene blue trihydrate 98.0-103.0% (dried basis), BAKER ANALYZED® C.I. 52015, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:7220-79-3
:
Formula:C₁₆H₁₈ClN₃S·3H₂O
MDL Number:MFCD00150008
:373.9 g/mol
:Management of Change (MOC) category = N
EINECS:200-515-2
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Solubility:Passes Test
- Identification:Passes Test
- Loss on Drying:8.0 - 18.0 %
- Residue after Ignition:≤1.2 %
- Copper (Cu):≤0.02 %
- Zinc (Zn):Passes Test
- Assay (C₁₆H₁₈ClN₃S) (dried basis):98.0 - 103.0 %
- Trace Impurities - Arsenic (As):≤8.000 ppm
Specification Test Results
Related Information
Certificates
Search by Lot NumberBrand Definition Baker Analyzed
Datasheet
Q473 Product Specifications
Additional Product Details
Frequently Bought Together