γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥99% (dry basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®
: 3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
:1132-61-2
:
:C₇H₁₅NO₄S
:MFCD00006183
:209.27 g/mol
:> 250 °C
:110 °C (closed cup)
:13,06288
:Management of Change (MOC) category = N
:214-478-5
:GHS07
:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Environmentally Preferable:
- Assay (C₇H₁₅NO₄S) (dried basis):≥99 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- Appearance (white to slightly pink powder to crystalline powder):Passes Test
- pH of 1% Solution at 25°C:2.5 - 4.5
- Solution (10%) in Water:Passes Test
- Loss on Drying:≤1 %
- Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm:≤0.020
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Frequently Bought Together