


Mercury (II) oxide, red ≥99.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:21908-53-2
:
Formula:HgO
MDL Number:MFCD00011045
:216.59 g/mol
Merck Index:13,05908
Boiling Pt:356.6 °C (1013 hPa)
Melting Pt:>400 °C
Density:11 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1641
ADR:6.1,II
:Management of Change (MOC) category = N
EINECS:244-654-7
:GHS06
:GHS08
:GHS09
Specification Test Results
- ACS - Assay (HgO):≥99.0 %
- ACS - Insoluble in Dilute HCl:≤0.03 %
- ACS - Residue after Reduction:≤0.025 %
- ACS - Chloride (Cl):≤0.025 %
- ACS - Sulfate (SO₄):≤0.015 %
- ACS - Nitrogen Compounds (as N):≤0.005 %
- ACS - Iron (Fe):≤0.005 %
Specification Test Results
Related Information
Certificates
Search by Lot Number2620 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
Frequently Bought Together