Orders LinkContactUsLinkComponent
Hydrometer Jars
Hydrometer Jars
Supplier:   Heathrow Scientific
Hydrometer Jars
Certificates
Hydrometer Jars
6 Options Available Below
Product Details & Documents
Hydrometer jars are made of borosilicate glass and feature a beaded rim with a circular base.
  • Available in a variety of capacities
All hydrometer jars are sized properly to eliminate surface tension effects that lead to inaccurate readings. Models 76546-096 have a graduation line at 1000 ml for precise filling and is designed for use as a sedimentation cylinder.
Product Family Options
Product Information
  • Capacity
    Use with
    Availability
    Price
  • 76546-092
    Without graduation lines
    Without graduation lines
    100 ml
    max. hydrometer length 185 mm (7.3")
    Log in for availability
    Log in for availability
    Each
    Each - $8.73
    $8.73
    New Customer Promotional Price
    Each
       $8.73
    Supplier Number: 21050-100
  • 76546-098
    Without graduation lines
    Without graduation lines
    250 ml
    max. hydrometer length 280 mm (11")
    Log in for availability
    Log in for availability
    Each
    Each - $12.50
    $12.50
    New Customer Promotional Price
    Each
       $12.50
    Supplier Number: 21050-250
  • 76546-100
    Without graduation lines
    Without graduation lines
    500 ml
    max. hydrometer length 360 mm (14.2")
    Log in for availability
    Log in for availability
    Each
    Each - $18.02
    $18.02
    New Customer Promotional Price
    Each
       $18.02
    Supplier Number: 21050-500
  • 76546-102
    Without graduation lines
    Without graduation lines
    600 ml
    max. hydrometer length 390 mm (15.4")
    Log in for availability
    Log in for availability
    Each
    Each - $18.66
    $18.66
    New Customer Promotional Price
    Each
       $18.66
    Supplier Number: 21050-600
  • 76546-094
    Without graduation lines
    Without graduation lines
    1000 ml
    max. hydrometer length 400 mm (15.7")
    Log in for availability
    Log in for availability
    Each
    Each - $29.65
    $29.65
    New Customer Promotional Price
    Each
       $29.65
    Supplier Number: 21050-1000
  • 76546-096
    With graduation lines
    With graduation lines
    1000 ml
    max. hydrometer length 400 mm (15.7")
    Log in for availability
    Log in for availability
    Each
    Each - $45.64
    $45.64
    New Customer Promotional Price
    Each
       $45.64
    Supplier Number: 21050-1000-L