

Iodine (iodine-iodide) 0.100 N (N/10)
Supplier: Ricca Chemical
Certificates
About this item
:
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0500 g/cm³
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear, dark brown, Iodine odor
- Assay at 20 °C (traceable to NIST SRM 136):0.1000 ± 0.0002 N at 20.0 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberRICCA Chemicals Ready-to-Use Titrants and Standards
Product Literature
Frequently Bought Together