



Iodine monochloride solution, StandARd®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
CAS Number:7790-99-0
:
Formula:ClI
MDL Number:MFCD00011354
:162.36 g/mol
ADR:8,3,II
:Management of Change (MOC) category = NI
UN:2920
:GHS02
:GHS05
:GHS08
:GHS07
Specification Test Results
- Assay (halogen content as Normality) (at time of packaging):0.215 - 0.225
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
H180 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together