KIMTECH PURE® G3 STERLING™ Nitrile Gloves, Ambidextrous, Kimberly-Clark
Supplier: Kimberly-Clark
Certificates
About this item
Powder-free, latex-free gloves provide the protection of nitrile and sensitivity comparable to natural rubber latex.
- Excellent tactile sensitivity with superior tensile strength
- Static-dissipative in use
Suitable for use in the manufacture of pharmaceuticals, medical devices, electronics, disk drives, flat panel displays, and semiconductors. Silicone-free. Ambidextrous. Beaded cuff. Textured fingertips. AQL 1.5. Packaged in a Class 100 cleanroom.
Certifications: Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higher cleanroom environments.
Specifications
- Catalog No:
- 89092-994
- Ambidextrous/Hand-specific:
- Ambidextrous
- Environmentally Preferable:
- Waste Reducing
- Powdered/Powder-free:
- Powder-Free
- Cuff Thickness:
- 2.8 mil
- Finger Thickness:
- 3.9 mil
- Palm Thickness:
- 3.1 mil