Order Entry
United States
ContactUsLinkComponent
n-Hexane ≥99.0% (by GC), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Flammable
Health
Irritant
Environment
n-Hexane ≥99.0% (by GC), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # JTN168-8
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Flammable
Health
Irritant
Environment
n-Hexane ≥99.0% (by GC), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # JTN168-8
Supplier Number:  N168-08
CAS Number:  
MOC:  Non-RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    4 L
  • Cat. No.:
    JTN168-8
  • Supplier no.:
    N168-08

Specifications

Specification Test Results

  • ACS - Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Assay (as n-Hexane) (by GC, corrected for water):
    ≥99.0 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤3 ppm
  • ACS - Water-Soluble Titrable Acid, meq/g:
    ≤0.0003
  • ACS - Sulfur Compounds (as S):
    ≤0.005 %
  • ACS - Thiophene:
    Passes Test
  • ACS - Water (by KF, coulometric):
    ≤0.03 %
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.3 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-280 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤0.04
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.20
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤0.30
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 195 nm:
    ≤1.00

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :Management of Change (MOC) category = N
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520