Order Entry
United States
ContactUsLinkComponent
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
Catalog # JTN151-5
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
Catalog # JTN151-5
Supplier Number:  N151-05
CAS Number:  
MOC:  Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    100 ml
  • Cat. No.:
    JTN151-5
  • Supplier no.:
    N151-05

Specifications

Specification Test Results

  • Assay (C₃H₂F₆0) (by GC, corrected for water):
    ≥99 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.100 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %

Specification Test Results

About this item

 :
Merck Index:13,09598
Boiling Pt:59.1 °C (1013 hPa)
Melting Pt:–96 °C
Density:1.492 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
ADR:8,II
 :Management of Change (MOC) category = N
 :168.04 g/mol
EINECS:213-059-4
UN:1760
 :GHS05
 :GHS07
CAS Number:920-66-1
Formula:C₃H₂F₆O
MDL Number:MFCD00011651