Order Entry
United States
ContactUsLinkComponent
Hydrochloric acid concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Corrosive
Irritant
Hydrochloric acid concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # JT4657-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Corrosive
Irritant
Hydrochloric acid concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # JT4657-1
Supplier Number:  4657-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    LDPE ampoule, DILUT-IT®
  • Size:
    1 pack
  • Cat. No.:
    JT4657-1
  • Supplier no.:
    4657-01

Specifications

Specification Test Results

  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
ADR:8,II
 :Management of Change (MOC) category = N
 :36.46
EINECS:231-595-7
UN:1789
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324