Cryogenic Tapes for Frozen Surfaces
Supplier: GA INTERNATIONAL INC
Certificates
About this item
These tapes are intended for identifying frozen containers and surfaces under deep freeze conditions. Ideal for long-term cryo preservation of samples in liquid nitrogen.
- Temperature range: −196 to 100 °C (−321 to 212 °F)
- Quickly adhere to already frozen containers and boxes
- Ideal for long-term storage in cryogenic and deep-freeze conditions
- Continuous format allows use of as much or little as necessary
- Available in a variety of colors for easy and convenient color-coding
Cryogenic tape for the identification of already frozen containers such as cryo boxes, cardboard and plastic freezer boxes, as well as tubes and bottles which are difficult to label under deep freeze conditions (−80 °C/−112 °F). Ideal for long-term cryo-preservation in liquid and vapor phase liquid nitrogen (−196 °C/−321 °F), laboratory freezers, and dry ice transportation. These cryo tapes can be inscribed using permanent cryo markers and are available in an assortment of colors for easy identification and color-coding.
Frequently Bought Together