L(+)-Glutamic acid ≥99.0% (dried basis, T), BAKER ANALYZED®, Biochemical Reagent
Catalog # JTM756-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: Non-RegulatedCertificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. No.:JTM756-7
- Supplier no.:M756-07
Specifications
Specification Test Results
- Assay (C₅H₉NO₄) (dried basis, by acid-base titrn):≥99.0 %
- Specific Rotation, [Å]²º^D(dried basis, c=2 in 5N HCl)(+):29.9 - 32.1 Degree
- Homogeneity by TLC:Passes Test
- Ash (sulfated):≤0.1 %
- Loss on Drying at 85°C:≤0.1 %
- Chloride (Cl):≤0.2 %
- Arsenic (As):≤0.0003 %
- Heavy Metals (as Pb):≤0.002 %
- Iron (Fe):≤0.003 %
Specification Test Results
About this item
CAS Number:56-86-0
:
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
:147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1.538 g/cm³ (20 °C)
:Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:200-293-7
MDL Number:MFCD00002634