Esteem® Stretchy Nitrile Ambi Gloves, Cardinal
Supplier: Avantor
Certificates
About this item
Ambidextrous synthetic nitrile gloves have beaded cuffs and textured fingers.
- Gloves are powder-and latex-free
Unique stretchy nitrile formulation provides the protection of nitrile while also offering comfort and flexibility.
Ordering information: Gloves are flat packed into double poly bags, heat-sealed, and placed into a poly case liner.
Frequently Bought Together