ContactUsLinkComponent
Finnpipette® Everyday Pipette and Microcentrifuge Bundle, Thermo Scientific
Finnpipette® Everyday Pipette and Microcentrifuge Bundle, Thermo Scientific
Catalog # 77392-034
undefined
Finnpipette® Everyday Pipette and Microcentrifuge Bundle, Thermo Scientific
Catalog # 77392-034
Supplier Number:  75003286
Specifications
  • Volume:
    0.2 - 2 µl, 2 - 20 µl, 20 - 200 µl, 100 - 1000 µl
  • Description:
    Everyday pipette and microcentrifuge bundle
  • Compatible with:
    Universal pipette tips
  • Cat. No.:
    77392-034
  • No. of channels:
    1
Specifications
About this item
Options
Product Information