Diethyl ether, anhydrous ≥99% (by GC, corrected for water content) stabilised, BAKER ANALYZED® A.C.S. Reagent ACS suitable for fat extraction, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane, Ethyl ether
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,I
:Management of Change (MOC) category = N
Flash Pt:–45 °C
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ≥7 ppm
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
:GHS02
:GHS07
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay ((C₂H₅)₂O) (by GC, corrected for water):≥99.0 %
- Alcohol (C₂H₅OH):Passes Test
- Carbonyl Compounds (as HCHO) (by polarography):≤0.001 %
- Color (APHA):≤10
- Peroxide (as H₂O₂):≤1 ppm
- Preservative (BHT):≥7 ppm
- Residue after Evaporation:≤0.001 %
- Titrable Acid (µeq/g):≤0.2
- Water (by KF, coulometric):≤0.01 %
Specification Test Results
Related Information
Certificates
Search by Lot Number9244 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Frequently Bought Together